missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEX15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565179
This item is not returnable.
View return policy
Description
Immunogen sequence: STQNNETELTS PILLPDLQIK ITNIFRPGFS PTADSLALKD SFCTHVTEAT KPEINKEDGE ILGFDIYSQP FGENADYPCE DKVDNIRQES GPVSNSEISL SFDLSR Highest antigen sequence identity to the following orthologs - mouse 50%, rat 48%.
Required during spermatogenesis for normal chromosome synapsis and meiotic recombination in germ cells. Necessary for formation of DMC1 and RAD51 foci on meiotic chromosomes, suggesting a specific role in DNA double-stranded break repair.
Specifications
| TEX15 | |
| Polyclonal | |
| Unconjugated | |
| TEX15 | |
| 2210014E14Rik; AL022622; AU022940; cancer/testis antigen 42; CT42; testis expressed 15; testis expressed gene 15; Testis-expressed protein 15; testis-expressed sequence 15 protein; TEX15 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56154 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BXT5 | |
| TEX15 | |
| Recombinant Human TEX15. Recombinant protein control fragment (Product #RP-105927). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction