missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEX38 Polyclonal Antibody

Description
This target displays homology in the following species: Human: 100%.
TEX38 is a protein coding gene.
Specifications
Specifications
| Antigen | TEX38 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | TEX38 |
| Gene Accession No. | Q6PEX7 |
| Gene Alias | 4930544O15Rik; ATPAF1 antisense gene protein 1; ATPAF1 antisense RNA 1; ATPAF1-AS1; C1orf223; testis expressed 38; testis highly expressed protein 4; Testis-expressed protein 38; testis-expressed sequence 38 protein; TEX38; THEG4 |
| Gene Symbols | TEX38 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?