Learn More
Texwipe™ IdealMat™ Contamination Control Mats - 30 Layer
Shop All Texwipe Products
Click to view available options
Color:
Blue
Gray
White
Dimensions (L x W):
25 x 45 in.
45 x 25 in.
46 x 36 in.
Unit Size:
Pack of 8
Description
Apply directly to floor
- Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
Specifications
Specifications
| Dimensions (L x W) | 46 x 36 in. |
| Quantity | 30 Sheets/Mat |
| Color | Blue |