Learn More
Invitrogen™ Tezepelumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559936
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Tezepelumab is under investigation in clinical trial NCT03406078 (Study to Evaluate the Efficacy and Safety of Tezepelumab in Reducing Oral Corticosteroid Use in Adults With Oral Corticosteroid Dependent Asthma).
Specifications
| Tezepelumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| AMG 157; AMG-157; MEDI9929; MEDI-9929 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG2 λ |
| ELISA, Flow Cytometry, Functional Assay | |
| 1 mg/mL | |
| 25mM histidine with 0.01% Tween 80, 8% sucrose and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |