missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFAP2E Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25829-1-AP-20UL
This item is not returnable.
View return policy
Description
TFAP2E is the sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP-2 factors bind to the consensus sequence 5′-GCCNNNGGC-3′ and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limb and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC. AP-2 epsilon may play a role in the development of the CNS and in cartilage differentiation.Specifications
| TFAP2E | |
| Polyclonal | |
| Unconjugated | |
| TFAP2E | |
| AP2 epsilon, AP2E, TFAP2E | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 313596, 332937, 339488 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6VUC0, Q6VUP9 | |
| Tfap2e | |
| TFAP2E Fusion Protein Ag23023 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction