missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFB1M Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 540.75
Specifications
| Antigen | TFB1M |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18467891
|
Novus Biologicals
NBP2-13429-25ul |
25ul |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18039409
|
Novus Biologicals
NBP2-13429 |
0.1 mL |
€ 572.00 € 540.75 / 0.10mL Save € 31.25 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TFB1M Polyclonal specifically detects TFB1M in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TFB1M | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| CGI75, CGI-75, dimethyladenosine transferase 1, mitochondrial, EC 2.1.1, EC 2.1.1.-, h-mtTFB, h-mtTFB1, homolog of yeast mitochondrial transcription factor B, hTFB1M, Mitochondrial 12S rRNA dimethylase 1, mitochondrial dimethyladenosine transferase 1, Mitochondrial transcription factor B1, mtTFB, mtTFB1, S-adenosylmethionine-6-N', N'-adenosyl(rRNA) dimethyltransferase 1, transcription factor B1, mitochondrial | |
| TFB1M | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Stem Cell Markers | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 51106 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: QRSRLSVMAQYLCNVRHIFTIPGQAFVPKPEVDVGVVHFTPLIQPKIEQPFKLVEKVVQNVFQFRRKYCHRGLRMLFPEAQRLESTGRLLELADIDPTLRPRQLSISHFKSLCDVYRKMCDEDPQL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title