missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFB1M Antibody (OTI6G4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74490FR
This item is not returnable.
View return policy
Description
TFB1M Monoclonal specifically detects TFB1M in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| TFB1M | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| TFB1M | |
| Recombinant protein fragment corresponding to amino acids 116-346 of human TFB1M (NP_057104) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI6G4 | |
| Western Blot | |
| CGI75, CGI-75, dimethyladenosine transferase 1, mitochondrial, EC 2.1.1, EC 2.1.1.-, h-mtTFB, h-mtTFB1, homolog of yeast mitochondrial transcription factor B, hTFB1M, Mitochondrial 12S rRNA dimethylase 1, mitochondrial dimethyladenosine transferase 1, Mitochondrial transcription factor B1, mtTFB, mtTFB1, S-adenosylmethionine-6-N′, N′-adenosyl(rRNA) dimethyltransferase 1, transcription factor B1, mitochondrial | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 51106.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction