missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFB1M Monoclonal Antibody (OTI11H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804100
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a dimethyltransferase that methylates the conserved stem loop of mitochondrial 12S rRNA. The encoded protein also is part of the basal mitochondrial transcription complex and is necessary for mitochondrial gene expression. The methylation and transcriptional activities of this protein are independent of one another. Variations in this gene may influence the severity of aminoglycoside-induced deafness.Specifications
| TFB1M | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WVM0 | |
| TFB1M | |
| Human recombit protein fragment corresponding to amino acids 116-346 of human TFB1M produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI11H1 | |
| Unconjugated | |
| TFB1M | |
| CGI-75, CGI75, mtTFB, mtTFB1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51106 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction