missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFB1M Monoclonal Antibody (OTI13H2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804108S
This item is not returnable.
View return policy
Description
TFB1M Monoclonal antibody specifically detects TFB1M in Human samples. It is validated for Western BlotSpecifications
| TFB1M | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14738 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5528 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI13H2 | |
| Unconjugated | |
| PPP2R5D | |
| CGI75; CGI-75; Dimethyladenosine transferase 1, mitochondrial; h-mtTFB; h-mtTFB1; homolog of yeast mitochondrial transcription factor B; hTFB1M; mitochondrial 12S rRNA dimethylase 1; mitochondrial dimethyladenosine transferase 1; Mitochondrial transcription factor B1; mtTFB; mtTFB1; OTTHUMP00000017473; S-adenosylmethionine-6-N', N'-adenosyl(rRNA) dimethyltransferase 1; TFB1M; transcription factor B1, mitochondrial | |
| Full length human recombinant protein of human PPP2R5D produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction