missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TFB1M Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519183
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
The protein encoded by this gene is a dimethyltransferase that methylates the conserved stem loop of mitochondrial 12S rRNA. The encoded protein also is part of the basal mitochondrial transcription complex and is necessary for mitochondrial gene expression. The methylation and transcriptional activities of this protein are independent of one another. Variations in this gene may influence the severity of aminoglycoside-induced deafness.
Specifications
| TFB1M | |
| Polyclonal | |
| Unconjugated | |
| TFB1M | |
| CGI75; CGI-75; Dimethyladenosine transferase 1, mitochondrial; h-mtTFB; h-mtTFB1; homolog of yeast mitochondrial transcription factor B; hTFB1M; mitochondrial 12S rRNA dimethylase 1; mitochondrial dimethyladenosine transferase 1; Mitochondrial transcription factor B1; mtTFB; mtTFB1; OTTHUMP00000017473; S-adenosylmethionine-6-N', N'-adenosyl(rRNA) dimethyltransferase 1; TFB1M; transcription factor B1, mitochondrial | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 51106 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8WVM0 | |
| TFB1M | |
| Synthetic peptide sequence (ELKRRKSKNEEKE) corresponding to the C-terminus amino acids of TFB1M (aa 325-338). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction