missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFDP2 Monoclonal Antibody (OTI4C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812077S
This item is not returnable.
View return policy
Description
TFDP2 Monoclonal antibody specifically detects TFDP2 in Human samples. It is validated for Western BlotSpecifications
| TFDP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75934 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10286 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4C11 | |
| Unconjugated | |
| BCAS2 | |
| 1110029I05Rik; A330080J22Rik; BTPS2; DP2; DP2.5; DP3; DP-3; E2F dimerization partner 2; GS; Tcfdp2; TFDP2; transcription factor Dp 2; transcription factor Dp-2; transcription factor Dp-2 (E2F dimerization partner 2); Unknown (protein for MGC:133813) | |
| Full length human recombinant protein of human BCAS2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction