missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TFE3 Polyclonal Antibody

Product Code. 15337624
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15337624 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15337624 Supplier Invitrogen™ Supplier No. PA535210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TFE3 Polyclonal Antibody for Western Blot, ICC/IF

The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. This protein contains a bipartite nuclear localization signal. This gene is known to escape chromosome X-inactivation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFE3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TFE3
Gene Accession No. P19532, Q64092
Gene Alias bHLHe33; Class E basic helix-loop-helix protein 33; F830016E06Rik; RCCP2; RCCX1; RGD1559642; Tcfe3; TFE3; Tfe-3; TFEA; transcription factor binding to IGHM enhancer 3; transcription factor E family, member A; transcription factor E3; transcription factor for IgH enhancer; transcription factor for immunoglobulin heavy-chain enhancer 3
Gene Symbols TFE3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human TFE3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209446, 7030
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.