missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFEB Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-12758IR
This item is not returnable.
View return policy
Description
TFEB Polyclonal specifically detects TFEB in Human samples. It is validated for Western Blot.
Specifications
| TFEB | |
| Polyclonal | |
| Western Blot | |
| ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 7942 | |
| Store at 4C in the dark. |
| Western Blot | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TFEB | |
| The immunogen this antibody was made to, maps to a region between residue 325 and 375 of human Transcription Factor EB using the numbering given in entry NP_009093.1 (GeneID 7942). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction