missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFEB Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41168H
This item is not returnable.
View return policy
Description
TFEB Polyclonal antibody specifically detects TFEB in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TFEB | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB | |
| Antibody was raised against a 16 amino acid peptide near the carboxy terminus of human TFEB. The immunogen is located within amino acids 390 - 440 of TFEB. Amino Acid Squence: FGGREDEGPPGYPEPL | |
| 0.1 mL | |
| Autophagy | |
| 7942 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction