missing translation for 'onlineSavingsMsg'
Learn More

TFEB Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications

Brand:  Novus Biologicals NB100-93447-20ug

 View more versions of this product

Product Code. 18509433

  • € 228.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TFEB Polyclonal specifically detects TFEB in Human, Rat samples. It is validated for Western Blot, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TFEB
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB
Goat
Affinity Purified
RUO
7942
Human, Rat
IgG
Western Blot, ELISA
0.5 mg/mL
Western Blot 1 - 3 μg/mL, Peptide ELISA Detection limit 1:1000
P19484
TFEB
Peptide with sequence C-QKDLQKSRELENH corresponding to internal region according to NP_009093.1.
20 μg
Primary
This antibody is expected not to cross-react to TFE3 and MITF.
Store at -20°C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only