missing translation for 'onlineSavingsMsg'
Learn More

TFEB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-12758

Product Code. 18779613

  • € 547.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TFEB Polyclonal specifically detects TFEB in Human samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TFEB
Polyclonal
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB
Rabbit
Affinity Purified
RUO
7942
Human
IgG
Western Blot, Immunoprecipitation
1.0 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 ug/mg lysate
NP_009093.1
TFEB
The immunogen this antibody was made to, maps to a region between residue 325 and 375 of human Transcription Factor EB using the numbering given in entry NP_009093.1 (GeneID 7942).
100 μL
Primary
Based on 100% sequence identity, this antibody is predicted to react with Goat, Bovine, Horse, Pig,Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Nine-banded armadillo, White-tufted-ear marmoset,Olive baboon and Northern white-cheeked gibbon.
Store at 4C. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

For Research Use Only