missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TFEB Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA596632

Product Code. 16529471

  • € 524.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Samples: Raji, NIH/3T3, mouse spleen, mouse kidney, rat brain, rat spleen; Cellular Location: Cytoplasm, Nucleus.

The Transcription factor EB (TFEB) is a member in the basic helix-loop-helix leucine zipper superfamily of transcription factors that is translocated in a subset of renal tumors. Recent studies have shown that lysosomal biogenesis is regulated by TFEB, which is in turn regulated by the mammalian target of rapamycin (mTOR) complex 1. Other evidence suggests that TFEB coordinates the major steps of the autophagic pathway by driving the expression of autophagy and lysosomal genes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TFEB
Polyclonal
Unconjugated
TFEB
Alpha TCFEB; ALPHATFEB; BHLHE35; Class E basic helix-loop-helix protein 35; RP4-696P19.3; T-cell transcription factor EB; Tcfeb; Tfeb; Transcription factor EB
Rabbit
Affinity Chromatography
RUO
21425, 316214, 7942
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
P19484, Q9R210
TFEB
Recombinant protein (or fragment).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.