missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TFEB Recombinant Rabbit Monoclonal Antibody (HL1065)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542385
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
The Transcription factor EB (TFEB) is a member in the basic helix-loop-helix leucine zipper superfamily of transcription factors that is translocated in a subset of renal tumors. Recent studies have shown that lysosomal biogenesis is regulated by TFEB, which is in turn regulated by the mammalian target of rapamycin (mTOR) complex 1. Other evidence suggests that TFEB coordinates the major steps of the autophagic pathway by driving the expression of autophagy and lysosomal genes.
Specifications
| TFEB | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P19484 | |
| TFEB | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human TFEB. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL1065 | |
| Unconjugated | |
| TFEB | |
| Alpha TCFEB; ALPHATFEB; BHLHE35; Class E basic helix-loop-helix protein 35; RP4-696P19.3; T-cell transcription factor EB; Tcfeb; Tfeb; Transcription factor EB | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7942 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction