missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFEC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13547-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the micropthalmia (MiT) family of basic helix-loop-helix leucine zipper transcription factors. MiT transcription factors regulate the expression of target genes by binding to E-box recognition sequences as homo- or heterodimers, and play roles in multiple cellular processes including survival, growth and differentiation. The encoded protein is a transcriptional activator of the nonmuscle myosin II heavy chain-A gene, and may also co-regulate target genes in osteoclasts as a heterodimer with micropthalmia-associated transcription factor. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
| TFEC | |
| Polyclonal | |
| Unconjugated | |
| TFEC | |
| bHLHe34, hTFEC L, TCFEC, TFE C, TFEC, TFECL, transcription factor EC, Transcription factor EC like | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21426, 22797, 26296 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14948, Q63302, Q9WTW4 | |
| Tfec | |
| TFEC Fusion Protein Ag4061 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion