missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TFF1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15925675
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15925675 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15925675 Supplier Invitrogen™ Supplier No. PA580108

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: mouse stomach tissue, mouse stomach tissue. IHC: mouse gaster tissue.

Members of the trefoil family are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. They are stable secretory proteins expressed in gastrointestinal mucosa. Their functions are not defined, but they may protect the mucosa from insults, stabilize the mucus layer, and affect healing of the epithelium. This gene, which is expressed in the gastric mucosa, has also been studied because of its expression in human tumors. This gene and two other related trefoil family member genes are found in a cluster on chromosome 21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFF1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene TFF1
Gene Accession No. Q08423
Gene Alias Bcei; breast cancer estrogen-inducible protein; breast cancer estrogen-inducible sequence; D21S21; gastrointestinal trefoil protein pS2; HP1.A; HPS2; PNR-2; polypeptide P1.A; Protein pS2; PS2; TFF1; Trefoil factor 1
Gene Symbols TFF1
Host Species Rabbit
Immunogen E. coli-derived mouse Tff1 recombinant protein (Position: Q22-F87).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21784
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.