missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFF1/pS2 Antibody (rTFF1/9134), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24121B
This item is not returnable.
View return policy
Description
TFF1/pS2 Monoclonal antibody specifically detects TFF1/pS2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TFF1/pS2 | |
| Monoclonal | |
| Biotin | |
| BCEIbreast cancer, estrogen-inducible sequence expressed in, Breast cancer estrogen-inducible protein, breast cancer estrogen-inducible sequence, D21S21, gastrointestinal trefoil protein pS2, hP1.A, HPS2, pNR-2, Polypeptide P1.A, Protein pS2, pS2, trefoil factor 1, trefoil factor, BCE1, human pS2 induced by estrogen from human breast cancercell line M10HP1.A | |
| Recombinant fragment (around aa1-84) of human TFF1/pS2 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Breast Cancer, Cancer | |
| 7031 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| rTFF1/9134 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction