missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFF1/pS2 Antibody (TFF1/2133), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08247AF350
This item is not returnable.
View return policy
Description
TFF1/pS2 Monoclonal antibody specifically detects TFF1/pS2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Specifications
| TFF1/pS2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| BCEIbreast cancer, estrogen-inducible sequence expressed in, Breast cancer estrogen-inducible protein, breast cancer estrogen-inducible sequence, D21S21, gastrointestinal trefoil protein pS2, hP1.A, HPS2, pNR-2, Polypeptide P1.A, Protein pS2, pS2, trefoil factor 1, trefoil factor, BCE1, human pS2 induced by estrogen from human breast cancercell line M10HP1.A | |
| Recombinant full-length human TFF1/pS2 protein (Uniprot: P04155) | |
| 100 μg | |
| Breast Cancer, Cancer | |
| 7031 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array | |
| TFF1/2133 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction