missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFF1/pS2 Mouse anti-Human, Clone: rTFF1/1091, Novus Biologicals™
Mouse Monoclonal Antibody
€ 499.00
Specifications
| Antigen | TFF1/pS2 |
|---|---|
| Clone | rTFF1/1091 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
TFF1/pS2 Monoclonal specifically detects TFF1/pS2 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| TFF1/pS2 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer | |
| 10 mM PBS with 0.05% BSA | |
| 7031 | |
| IgG1 κ | |
| Protein A or G purified |
| rTFF1/1091 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BCEIbreast cancer, estrogen-inducible sequence expressed in, Breast cancer estrogen-inducible protein, breast cancer estrogen-inducible sequence, D21S21, gastrointestinal trefoil protein pS2, hP1.A, HPS2, pNR-2, Polypeptide P1.A, Protein pS2, pS2, trefoil factor 1, trefoil factor, BCE1, human pS2 induced by estrogen from human breast cancercell line M10HP1.A | |
| Recombinant full-length human TFF1/pS2 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title