missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFF3 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76514AF700
This item is not returnable.
View return policy
Description
TFF3 Polyclonal antibody specifically detects TFF3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| TFF3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| HITF, human intestinal trefoil factor10Intestinal trefoil factor, trefoil factor 3 (intestinal) | |
| Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human TFF3. The immunogen is located within the last 50 amino acids of TFF3. Amino Acid Squence: ECNNRGCCFDSRIPGV | |
| 0.1 mL | |
| Biologically Active Proteins, Cancer | |
| 7033 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction