missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFF3 Antibody (SR1054), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 449.00
Specifications
| Antigen | TFF3 |
|---|---|
| Clone | SR1054 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663734
|
Novus Biologicals
NBP3-22003-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669674
|
Novus Biologicals
NBP3-22003-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TFF3 Monoclonal antibody specifically detects TFF3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
| TFF3 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| HITF, human intestinal trefoil factor10Intestinal trefoil factor, trefoil factor 3 (intestinal) | |
| A synthesized peptide derived from human TFF3 (Uniprot #: Q07654) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1054 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins, Cancer | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 7033 | |
| IgG | |
| Affinity purified |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto