missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFG Antibody (OTI2C3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-01438
This item is not returnable.
View return policy
Description
TFG Monoclonal specifically detects TFG in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TFG | |
| Monoclonal | |
| 0.45 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
| TFG | |
| Full length human recombinant protein of human TFG(NP_001007566) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI2C3 | |
| Unconjugated | |
| FLJ36137, protein TFG, TF6, TRK-fused gene, TRK-fused gene protein, TRKT3, TRKT3 oncogene | |
| Mouse | |
| 43.3 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 10342 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction