missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFIIB Antibody (OTI1H10), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74492AF647
This item is not returnable.
View return policy
Description
TFIIB Monoclonal specifically detects TFIIB in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| TFIIB | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GTF2B | |
| Recombinant protein fragment corresponding to amino acids 43-316 of human TFIIB (NP_001505) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI1H10 | |
| Western Blot | |
| general transcription factor IIB, General transcription factor TFIIB, S300-II, TFIIBTF2BRNA polymerase II transcription factor IIB, transcription initiation factor IIB | |
| Mouse | |
| Affinity Purified | |
| DNA replication Transcription Translation and Splicing | |
| 2959.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction