missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFIIB Antibody (SR1524), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22207-100ul
This item is not returnable.
View return policy
Description
TFIIB Monoclonal antibody specifically detects TFIIB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| TFIIB | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR1524 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| general transcription factor IIB, General transcription factor TFIIB, S300-II, TFIIBTF2BRNA polymerase II transcription factor IIB, transcription initiation factor IIB | |
| A synthesized peptide derived from human TFIIB (Uniprot #: Q00403) | |
| 100 μg | |
| DNA replication Transcription Translation and Splicing | |
| 2959 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction