missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFIIE-alpha Antibody (SR2046), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | TFIIE-alpha |
|---|---|
| Clone | SR2046 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry, Serum 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18634315
|
Novus Biologicals
NBP3-22636-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656454
|
Novus Biologicals
NBP3-22636-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TFIIE-alpha Monoclonal antibody specifically detects TFIIE-alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, SerumSpecifications
| TFIIE-alpha | |
| Western Blot 1:500-1:2000, Immunohistochemistry, Serum 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FE, General transcription factor IIE 56 kDa subunit, general transcription factor IIE subunit 1, general transcription factor IIE, polypeptide 1 (alpha subunit, 56kD), general transcription factor IIE, polypeptide 1, alpha 56kDa, TF2E1, TFIIE-A, TFIIE-alpha, Transcription initiation factor IIE subunit alpha | |
| A synthesized peptide derived from human TFIIE-alpha (Uniprot #: P29083) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2046 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 2960 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title