missing translation for 'onlineSavingsMsg'
Learn More

TFIP11 Antibody (3H5) [Alexa Fluor« 750], Novus Biologicals Biologicals™

Product Code. 30522288 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30522288 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30522288 Supplier Novus Biologicals Supplier No. NBP343044AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TFIP11 Monoclonal antibody specifically detects TFIP11 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFIP11
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3H5
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias bK445C9.6, DKFZP434B194, FLJ22086, hNtr1, NTR1, Septin and tuftelin-interacting protein 1, STIP, STIP-1, TIP39Spp382, tuftelin interacting protein 11, tuftelin-interacting protein 11
Host Species Mouse
Immunogen tuftelin interacting protein 11 - (Uniprot# Q9UBB9)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24144
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.