missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TFM-4AS-1
GREENER_CHOICE

Product Code. 18005692
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18005692

Brand: Tocris Bioscience™ 3813/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective androgen receptor modulator (SARM)

Potent selective androgen receptor modulator (SARM) (IC50 = 30 nM). Steroidal compound. Exhibits limited effects on reproductive tissues and sebaceous glands; does not repress AP-1-sensitive MMP-1 reporter. Inhibits 5α-reductase type I and II (IC50 values are 2 and 3 nM respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188589-61-9
Molecular Formula C27H33F3N2O2

Specifications

Quantity 50 mg
Formula Weight 474.56
Purity >98%
Molecular Weight (g/mol) 474.568
Chemical Name or Material TFM-4AS-1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.