missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFPI Antibody (401), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89570PCP
This item is not returnable.
View return policy
Description
TFPI Monoclonal antibody specifically detects TFPI in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ELISA
Specifications
| TFPI | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 401 | |
| Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA Detection | |
| convertin, EPITFPI1LACITFI, Extrinsic pathway inhibitor, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human TFPI (Accession#: NP_006278.1; Met1-Lys282). | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 7035 | |
| Store at 4C in the dark. | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering