missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFPI Antibody (6), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89569MFV500
This item is not returnable.
View return policy
Description
TFPI Monoclonal antibody specifically detects TFPI in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| TFPI | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 6 | |
| Western Blot, Immunoprecipitation | |
| convertin, EPITFPI1LACITFI, Extrinsic pathway inhibitor, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human TFPI (Accession#: NP_006278.1; Met 1-Lys 282). | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 7035 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction