missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFPI Antibody (JE40-58), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33007
This item is not returnable.
View return policy
Description
TFPI Monoclonal antibody specifically detects TFPI in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| TFPI | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry, Immunoprecipitation 1-2ug/sample, Immunohistochemistry-Paraffin 1:200 | |
| convertin, EPITFPI1LACITFI, Extrinsic pathway inhibitor, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) | |
| Synthetic peptide within Human TFPI aa 250-304. (Uniprot: P10646) | |
| 100 μL | |
| Cardiovascular Biology | |
| 7035 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| JE40-58 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction