missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFPI Antibody (OTI5H5), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74493V
This item is not returnable.
View return policy
Description
TFPI Monoclonal specifically detects TFPI in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TFPI | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TFPI | |
| Recombinant protein fragment corresponding to amino acids 29-304 of human TFPI (NP_006278) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5H5 | |
| Western Blot, Immunohistochemistry | |
| convertin, EPITFPI1LACITFI, Extrinsic pathway inhibitor, Lipoprotein-associated coagulation inhibitor, tissue factor pathway inhibitor, tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor) | |
| Mouse | |
| Affinity Purified | |
| Cardiovascular Biology | |
| 7035.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction