Learn More
TFPI2 Mouse anti-Human, Clone: OTI1H10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF805268
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TFPI2 may play a role in the regulation of plasmin-mediated matrix remodeling. It inhibits trypsin, plasmin, factor VIIa/tissue factor and weakly factor Xa. TFPI2 has no effect on thrombin.Specifications
| TFPI2 | |
| Monoclonal | |
| Unconjugated | |
| TFPI2 | |
| PP5, REF1, TFPI-2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7980 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1H10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P48307 | |
| TFPI2 | |
| Human recombit protein fragment corresponding to amino acids 23-235 of human TFPI2 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.