Learn More
Invitrogen™ TFPI2 Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA547343
Product Code. 15577832
  • € 520.00 / 100µg
Description
In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse TFPI-2 is observed and less than 5% cross-reactivity with recombinant human TFPI-1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
TFPI2 may play a role in the regulation of plasmin-mediated matrix remodeling. It inhibits trypsin, plasmin, factor VIIa/tissue factor and weakly factor Xa. TFPI2 has no effect on thrombin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TFPI2
Polyclonal
Unconjugated
TFPI2
AV000670; Placental protein 5; PP5; PP5/TFPI-2; PP5FLJ21164; REF1; REF-1; retinal pigment epithelium cell factor 1; TFPI2; TFPI-2; TFPI-2REF1; Tissue factor pathway inhibitor 2
Goat
Antigen affinity chromatography
RUO
7980
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Immunoprecipitation, Neutralization, Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
P48307
TFPI2
Mouse myeloma cell line NS0-derived recombinant human TFPI-2 Asp23-Lys213.