missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFPT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10097-2-AP-150UL
This item is not returnable.
View return policy
Description
TFPT component of the chromatin-remodeling INO80 complex which, in addition to chromatin remodeling implicated in crucial functions such as DNA repair, checkpoint regulation, DNA replication, telomemere maintece and chromosome segregation. This protein appears to promote apoptosis in a TP53/p53-independent manner.Specifications
| TFPT | |
| Polyclonal | |
| Unconjugated | |
| TFPT | |
| amida, FB1, INO80 complex subunit F, INO80F, Protein FB1, TCF3 fusion partner, TFPT | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29844, 69714, 85423 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P0C1Z6, Q3U1J1, Q9JMG6 | |
| Tfpt | |
| TFPT Fusion Protein Ag0131 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction