missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TfR2 Antibody (353810) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB31202JF669
This item is not returnable.
View return policy
Description
TfR2 Monoclonal antibody specifically detects TfR2 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| TfR2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| HFE3transferrin receptor protein 2, MGC126368, TfR2, TFRC2, transferrin receptor 2 | |
| Mouse myeloma cell line NS0-derived recombinant human TfR2, Arg105-Phe801, Accession # Q9UP52 | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 7036 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Western Blot, CyTOF | |
| 353810 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction