Learn More
TfR2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-93436-0.1ml
 View more versions of this product
Product Code. 18693161
  • € 506.00 / 0.10mL
Description
TfR2 Polyclonal antibody specifically detects TfR2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TfR2
Polyclonal
Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
HFE3transferrin receptor protein 2, MGC126368, TfR2, TFRC2, transferrin receptor 2
Recombinant fusion protein containing a sequence corresponding to amino acids 130-330 of human TFR2 (NP_003218.2). FHQGRLYWSDLQAMFLQFLGEGRLEDTIRQTSLRERVAGSAGMAALTQDIRAALSRQKLDHVWTDTHYVGLQFPDPAHPNTLHWVDEAGKVGEQLPLEDPDVYCPYSAIGNVTGELVYAHYGRPEDLQDLRARGVDPVGRLLLVRVGVISFAQKVTNAQDFGAQGVLIYPEPADFSQDPPKPSLSSQQAVYGHVHLGTGDP
0.1 mL
Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
7036
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse