missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOSTRIN Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94431-0.1ml
This item is not returnable.
View return policy
Description
NOSTRIN Polyclonal antibody specifically detects NOSTRIN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| NOSTRIN | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| BM247 homolog, DaIP2, endothelial nitric oxide synthase traffic inducer, ENOS traffic inducer, eNOS-trafficking inducer, Nitric oxide synthase traffic inducer, nitric oxide synthase traffickerMGC20702, nitric oxide synthase trafficking, nostrin, ortholog of mouse disabled 2 interacting proein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 270-430 of human NOSTRIN (NP_001034813.2). ILSTENKSEFLLTDYFEEDPNSAMDKERRKSLLKPKLLRLQRDIEKASKDKEGLERMLKTYSSTSSFSDAKSQKDTAALMDENNLKLDLLEANSYKLSSMLAELEQRPQPSHPCSNSIFRWREKEHTHSYVKISRPFLMKRLENIVSKASSGGQSNPGSST | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 115677 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction