missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TFR2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17884770
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17884770 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17884770 Supplier Invitrogen™ Supplier No. PA5140257

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: SVLVVSEDVN YEPDLDFHQG RLYWSDLQAM FLQFLGEGRL EDTIRQTSLR ERVAGSAGMA ALTQDIRAAL SRQKLDHVWT DTHYVGLQFP DPAHPNTLHW VDEAGKVGEQ LPLEDPDVYC PYS Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 80%.

This gene is a member of the transferrin receptor-like family and encodes a single-pass type II membrane protein with a protease associated (PA) domain, an M28 peptidase domain and a transferrin receptor-like dimerization domain. This protein mediates cellular uptake of transferrin-bound iron and mutations in this gene have been associated with hereditary hemochromatosis type III. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFR2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tfr2
Gene Accession No. Q9UP52
Gene Alias HFE3; MGC126368; TfR2; TFRC2; transferrin receptor 2; transferrin receptor protein 2; Trfr2
Gene Symbols Tfr2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TFR2. Recombinant protein control fragment (Product #RP-109320).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7036
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.