missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TG Monoclonal Antibody (OTI15F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804642S
This item is not returnable.
View return policy
Description
TG Monoclonal antibody specifically detects TG in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| TG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UMY4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29934 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI15F1 | |
| Unconjugated | |
| SNX12 | |
| AITD3; cog; Colloid; congenital goiter; hereditary goitre; TG; Tgn; thyroglobulin; thyroglobulin precursor | |
| Full length human recombinant protein of human SNX12 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction