missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TG Monoclonal Antibody (OTI8F4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804209S
This item is not returnable.
View return policy
Description
TG Monoclonal antibody specifically detects TG in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| TG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P08151 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2735 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI8F4 | |
| Unconjugated | |
| GLI1 | |
| AITD3; cog; Colloid; congenital goiter; hereditary goitre; TG; Tgn; thyroglobulin; thyroglobulin precursor | |
| Human recombinant protein fragment corresponding to amino acids 755-1106 of human GLI1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction