missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF beta 1/2/3 Antibody (1D11) [Janelia Fluor™ 669], Novus Biologicals™
Shop All Bio Techne ProductsDescription
TGF beta 1/2/3 Monoclonal antibody specifically detects TGF beta 1/2/3 in Broad Species Reactivity samples. It is validated for Western Blot,Binding Inhibition,ELISA Standard (Matched Pair),Neutralization,ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
Specifications
Specifications
| Antigen | TGF beta 1/2/3 |
| Applications | Western Blot, Binding Inhibition Assay, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) |
| Classification | Monoclonal |
| Clone | 1D11 |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor, beta 1 |
| Host Species | Mouse |
| Immunogen | Bovine bone-derived TGF-β1 and TGF-β2 |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?