missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 1 Antibody (MM0446-6G14), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11766G
This item is not returnable.
View return policy
Description
TGF-beta 1 Monoclonal specifically detects TGF-beta 1 in Human samples. It is validated for Flow Cytometry.
Specifications
| TGF-beta 1 | |
| Monoclonal | |
| DyLight 488 | |
| P01137 | |
| TGFB1 | |
| Human recombinant TGF-beta 1 | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific | |
| 7040 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry | |
| MM0446-6G14 | |
| Western Blot, Flow Cytometry | |
| CEDLAP, DPD1, latency-associated peptide, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction