missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 1 Antibody (OTI1E12), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74495F
This item is not returnable.
View return policy
Description
TGF-beta 1 Monoclonal specifically detects TGF-beta 1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| TGF-beta 1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| TGFB1 | |
| Full length recombinant protein of human TGF-beta 1 (NP_000651) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI1E12 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| CEDLAP, DPD1, latency-associated peptide, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
| Mouse | |
| Affinity Purified | |
| Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific, Signal Transduction | |
| 7040 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction