missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 1 Antibody (SR1847), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | TGF-beta 1 |
|---|---|
| Clone | SR1847 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600954
|
Novus Biologicals
NBP3-21548-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609424
|
Novus Biologicals
NBP3-21548-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TGF-beta 1 Monoclonal antibody specifically detects TGF-beta 1 in Human samples. It is validated for Western BlotSpecifications
| TGF-beta 1 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CEDLAP, DPD1, latency-associated peptide, TGF beta, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
| A synthesized peptide derived from human TGF-beta 1 (Uniprot #: P01137) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| SR1847 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 7040 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title