missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TGF beta-1 Recombinant Rabbit Monoclonal Antibody (PD00-17)
GREENER_CHOICE

Product Code. 17989402
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17989402 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17989402 Supplier Invitrogen™ Supplier No. MA550685

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive control: Raji cell lysate, K-562 cell lysate, mouse spleen tissue lysate, rat spleen tissue lysate, rat spleen tissue, mouse spleen tissue. Predicted band size: 44 kDa Subcellular Location: Extracellular matrix, Secreted.

TGF beta-1 (TGFB1, Transforming Growth Factor Beta 1) is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its pro-domain. Activation of TGF beta-1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF beta-1, an amino-terminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP. TGF beta-1 helps regulates proliferation, differentiation, adhesion, migration in many cell types. Many cells have TGF beta receptors, and the protein positively and negatively regulates many other growth factors. TGF beta-1 is cleaved into a latency-associated peptide and a mature TGF beta-1 peptide, and is found in either a latent form composed of a TGFB1 homodimer, a LAP homodimer, and a latent TGFB1-binding protein, or in an active form composed of a TGF beta-1 homodimer. The mature peptide may also form heterodimers with other TGF beta family members. The gene for TGF beta-1 is frequently upregulated in tumor cells, and mutations in this gene result in Camurati-Engelmann disease and cystic fibrosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TGF beta-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone PD00-17
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Tgfb1
Gene Accession No. P01137, P04202, P17246
Gene Alias CED; cytokine; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGF b; TGF b 1; TGF beta; tgf beta 1; tgf beta1; TGF beta-1; TGF beta-1 protein; TGF β; TGF β 1; TGF β1; Tgfb; TGFB1; TGF-B1; Tgfb-1; TGFbeta; TGF-beta 1; TGF-beta 1 protein; TGFbeta1; TGF-beta1; TGF-beta-1; TGFβ1; Transforming growth factor; transforming growth factor b1; transforming growth factor beta; transforming growth factor beta 1; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor beta-1 proprotein; transforming growth factor beta-1; transforming growth factor β1; transforming growth factor, beta 1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1; transforming growth factor-beta I; transforming growth factor-beta-1; transforming growth factor-beta-1 precursor; unnamed protein product
Gene Symbols Tgfb1
Host Species Rabbit
Immunogen Synthetic peptide (KLH-coupled) corresponding to Human TGF beta-1 C-terminal.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21803, 59086, 7040
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.