missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 2 Antibody (MM0640-6M28), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32727AF488
This item is not returnable.
View return policy
Description
TGF-beta 2 Monoclonal specifically detects TGF-beta 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TGF-beta 2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TGFB2 | |
| Human recombinant TGF-beta 2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0640-6M28 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| BSC-1 cell growth inhibitor, Cetermin, Glioblastoma-derived T-cell suppressor factor, G-TSF, MGC116892, Polyergin, TGF-beta2, TGF-beta-2, transforming growth factor beta-2, transforming growth factor, beta 2 | |
| Mouse | |
| Protein G purified | |
| Angiogenesis, Apoptosis, Biologically Active Proteins, Cell Cycle and Replication | |
| 7042 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction